Spex WP-15 Assurance® Grade Trace Metals with Mercury in 5% HNO3; 125 mL

Representative image only.

Spex WP-15 Assurance® Grade Trace Metals with Mercury in 5% HNO3; 125 mL

Spex | Item: 0456841

Multi-element standards for AA and ICP analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • May be used either as standards or as a means to check the individual analysts accuracy and precision
  • Trace metals standards with mercury in 5% HNO3 matrix
  • 125 mL bottle

More About this Item

Standards may be diluted in the same matrix as specified; however, caution must be exercised in the choice of the source for your diluents. Diluting the matrix may cause some standards to precipitate. Also, an impure or unknown diluent turns your standard into an unknown. We recommend using only SPEX CertiPrep Matrix Blanks when diluting your standards.

Refer to US EPA Methods Manual 600/4-79-020 "Methods for Chemical Analysis of Water and Wastes" Trace Metals 21I, 21II and 21III Methods.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-element standard contains 15 elements.

• Trace metals with mercury standard is a 125 mL bottle of 500 µg/mL (500 ppm) Aluminum; 250 µg/mL (250 ppm) Vanadium; 100 µg/mL (100 ppm) Arsenic, Beryllium, Chromium, Cobalt, Copper, Iron, Lead, Manganese, Nickel, Zinc; 25 µg/mL Cadmium, Selenium; and 10 µg/mL (10 ppm) Mercury* in 5% HNO3.

Hazardous material shipping fees may apply.

*Mercury is supplied as a separate solution (04503-58) due to incompatibility with other elements.

Specifications
  • ConcentrationMultiple
  • Purity GradeAssurance® Grade
  • Matrix5% HNO3
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionAssurance® Grade Trace Metals with Mercury for AA & ICP in 5% HNO3; 125 mL
Documents & Certificates
Reviews
Q&A