Spex WP-11-500 Assurance® Grade Alternate Metals for AA & ICP in 2% HNO3; 500 mL

Representative image only.

Spex WP-11-500 Assurance® Grade Alternate Metals for AA & ICP in 2% HNO3; 500 mL

Spex | Item: 0456850

Multi-element groundwater and wastewater pollution control check standards for AA & ICP analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • Refer to US EPA Methods Manual 600/4-79-020 "Methods for Chemical Analysis of Water and Wastes" Trace Metals 21I, 21II and 21III Methods
  • Alternate metals standard for AA & ICP in 2% HNO3 matrix
  • 500 mL bottle

More About this Item

May be used either as standards or as a means to check the individual analysts accuracy and precision.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-element standard contains 11 elements.

• Alternate Metals Standard is a 500 mL bottle which contains 20 µg/mL (20 ppm) of Aluminum, Iron, Vanadium; 10 µg/mL (10 ppm) of Cobalt, Copper, Manganese, Nickel, Zinc; and 5 µg/mL (5 ppm) of Antimony, Beryllium, Thallium.

Hazardous material shipping fees may apply.

Specifications
  • Purity GradeAssurance® Grade
  • ConcentrationMultiple
  • Matrix2% HNO3
  • Element/IonAluminum, Antimony, Beryllium, Cobalt, Copper, Iron, Manganese, Nickel, Thallium, Vanadium, and Zinc
  • FormLiquid
  • Volume (mL)500
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Grup III
  • Storage SymbolAmbient
  • Hazard SymbolIrritant
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionSpex CertiPrep Assurance® Grade Alternate Metals for AA & ICP in 2% HNO3; 500 mL
Documents & Certificates
Reviews
Q&A