Spex VIN-AC Vinyl Acetate Organic Standard, 2,000 µg/mL in Methanol-P&T; 1 mL

Representative image only.

Spex VIN-AC Vinyl Acetate Organic Standard, 2,000 µg/mL in Methanol-P&T; 1 mL

Spex | Item: 0456647

Analytical standards for drinking water, wastewater, and solid waste

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Methods 524.2, 624, and 8260
  • 2,000 ppm vinyl acetate organic standard in methanol-P&T matrix
  • 1 mL ampule

More About this Item

US EPA Method 524.2 is a general analytical method for the measurement of purgeable organic compounds in water by capillary column gas chromatography/mass spectrometry.

US EPA Method 624 is a general analytical method for the monitoring of VOCs in municipal and industrial wastewater by GC/MS.

US EPA Method 8260 is a general analytical method for the monitoring of VOCs by GC/MS.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

Specifications
  • Concentration2,000 µg/mL (2,000 ppm)
  • MatrixMethanol-P&T
  • Element/IonVinyl acetate
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Methanol; Class 3, Packing Group II
  • Storage SymbolRefrigerator
  • Hazard SymbolCorrosion, Irritant, Health Hazard
  • CAS Number108-05-4
  • UN NumberUN1230
  • Shelf Life (months)12
  • DescriptionVinyl Acetate Organic Standard, 2,000 µg/mL in Methanol-P&T; 1 mL
Documents & Certificates
Reviews
Q&A