Spex USPS2380-DMSO-PK Methanol USP <467> Residual Solvent Standard, 15,000 µg/mL; 1 mL; 4/pk

Representative image only.

Spex USPS2380-DMSO-PK Methanol USP <467> Residual Solvent Standard, 15,000 µg/mL; 1 mL; 4/pk

Spex | Item: 0500951

Comply with the guidelines set by the USP and the ICH to identify and quantify residual solvents

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 15,000 ppm methanol USP <467> residual solvent standard in dimethyl sulfoxide
  • Four 1 mL ampules

More About this Item

In the pharmaceutical industry, the guidelines set by the International Conference on Harmonization (ICH), and by United States Pharmacopeia (USP) and European Pharmacopeia (EP), mandate that manufacturing solvents have to be regulated due to their toxic and/or environmentally hazardous nature.

The USP General Chapter <467> Residual Solvents is a method widely used for identifying and quantifying residual solvents when there is no information available on what solvents are likely to be present. Our products are manufactured and verified by our QC department to validate the Certificate of Analysis which accompanies each product. Spex CertiPrep® offers analytes in dimethyl sulfoxide for Class 1, Class 2 and Class 3 solvents.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration2,000 µg/mL (2,000 ppm)
  • ChemistryOrganics;Standards
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)1
  • Packaging4 x 1 mL
  • DOT InformationHazardous; Acetone; Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Irritant, Health Hazard
  • UN NumberUN1090
  • CAS Number71-55-6, 513-88-2, 96-12-8, 106-93-4, 75-27-4, 75-25-2, 56-23-5, 67-66-3, 76-06-2, 3252-43-5, 124-48-1, 3018-12-0, 127-18-4, 545-06-2, 79-01-6
  • Shelf Life (months)12
  • DescriptionChlorinated Disinfectant By-Products Mix for US EPA Method 551, 2,000 µg/mL in Acetone; 1 mL
Documents & Certificates
Reviews
Q&A