Spex USP-TXM2A Oral Elemental Impurities A in 5% HNO3/1% HCl; 125 mL

Representative image only.

Spex USP-TXM2A Oral Elemental Impurities A in 5% HNO3/1% HCl; 125 mL

Spex | Item: 0456960

Impurities mix helps meet the new limits in pharmaceutical products for arsenic, cadmium, lead, and mercury

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Use with ICP-MS instrumentation for the analysis of low level impurities
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • Oral elemental impurities A in 5% HNO3/1% HCl matrix
  • 125 mL bottle

More About this Item

The new guidelines set by the United States Pharmacopeia (USP) have pushed the pharmaceutical and nutraceutical industries to provide accurate, quantifiable results for metal analysis in drugs, pharmaceutical substances, and raw materials.

USP <232> outlines new limits in pharmaceutical products for arsenic, cadmium, lead, and mercury. The procedures focus on the use of ICP-MS for the analysis of low level impurities. ICP-MS instrumentation, along with accurate ICP-MS standards, allow for increased efficiency and accuracy of the analysis necessary to comply with the new regulations.

Use these standards as a calibration or check standard to verify Oral Daily Dose PDE, Parenteral Component Limit, or Parenteral Daily Dose PDE as well as Inhalation Component Limit or Daily Dose.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component standard contains 4 components.

• Oral Elemental Impurities A is a 125 mL bottle which contains 30 mg/kg of Mercury, 15 mg/kg of Arsenic, and 5 mg/kg of Cadmium and Lead.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Matrix5% HNO3/1% HCl
  • Element/IonArsenic, Cadmium, Lead, and Mercury
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN3264
  • Shelf Life (months)6
  • DescriptionOral Elemental Impurities A in 5% HNO3/1% HCl; 125 mL
Documents & Certificates
Reviews
Q&A