Spex USP-RS-C3A 24 Organic Volatile Impurities, USP <467> Pharmaceutical Residual Solvent; 1 mL

Representative image only.

Spex USP-RS-C3A 24 Organic Volatile Impurities, USP <467> Pharmaceutical Residual Solvent; 1 mL

Spex | Item: 0455986

Comply with the guidelines set by the USP and the ICH to identify and quantify residual solvents

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • USP <467> pharmaceutical residual solvent standard in dimethyl sulfoxide matrix
  • Class 3 solvents
  • 1 mL ampule

More About this Item

In the pharmaceutical industry, the guidelines set by the International Conference on Harmonization (ICH), and by United States Pharmacopeia (USP) and European Pharmacopeia (EP), mandate that manufacturing solvents have to be regulated due to their toxic and/or environmentally hazardous nature.

The USP General Chapter <467> Residual Solvents is a method widely used for identifying and quantifying residual solvents when there is no information available on what solvents are likely to be present. Our products are manufactured and verified by our QC department to validate the Certificate of Analysis which accompanies each product. Spex CertiPrep® offers analytes in dimethyl sulfoxide for Class 1, Class 2 and Class 3 solvents.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component solution contains 24 components.

• 24 Organic Volatile Impurities, Pharmaceutical Residual Solvent Standard is a 1 mL ampule which contains 1,000 µg/mL (1,000 ppm) of Acetone, Anisole, 1-Butanol, 2-Butanol, 2-Butanone, Butyl acetate, methyl tertiary-Butyl ether, Ethanol, Ether, Ethyl acetate, Ethyl formate, n-Heptane, Isobutyl acetate, Isopropyl acetate, Isopropylbenzene, Methyl acetate, 3-Methyl-1-butanol, 4-Methyl-2-pentanone, 2-Methyl-1-propanol, n-Pentane, 1-Pentanol, 1-Propanol, 2-Propanol, and Propyl acetate.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixDimethyl Sulfoxide
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)1
  • DOT InformationNot Regulated
  • Storage SymbolFreezer
  • Hazard SymbolIrritant, Health Hazard
  • CAS Number67-64-1, 100-66-3, 71-36-3, 78-92-2, 78-93-3, 123-86-4, 1634-04-4, 64-17-5, 60-29-7, 141-78-6, 109-94-4, 142-82-5, 110-19-0, 108-21-4, 98-82-8, 79-20-9, 123-51-3, 108-10-1, 78-83-1, 109-66-0, 71-41-0, 71-23-8, 67-63-0, 109-60-4
  • UN NumberNot Regulated
  • Shelf Life (months)3
  • Description24 Organic Volatile Impurities, Pharmaceutical Residual Solvent Standards in Dimethyl Sulfoxide; 1 mL
Documents & Certificates
Reviews
Q&A