Spex USP-PARENT3 USP <232> Revision 40, Parenteral Mix 3 Elemental Impurities; 125 mL

Representative image only.

Spex USP-PARENT3 USP <232> Revision 40, Parenteral Mix 3 Elemental Impurities; 125 mL

Spex | Item: 0456953

Use with ICP-MS instrumentation for the analysis of low level impurities

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 & ISO 17034:2016
  • Certified by DQS to ISO 9001:2015
  • Helps meet the new limits in pharmaceutical products for arsenic, cadmium, lead, and mercury
  • USP <232> Revision 40, Parenteral Mix 3 elemental impurities in 5% HNO3/tr. Tartaric Acid/tr. HF matrix
  • 125 mL bottle

More About this Item

The new guidelines set by the United States Pharmacopeia (USP) have pushed the pharmaceutical and nutraceutical industries to provide accurate, quantifiable results for metal analysis in drugs, pharmaceutical substances and raw materials.

USP <232> outlines new limits in pharmaceutical products for arsenic, cadmium, lead, and mercury. ICP-MS instrumentation, along with accurate ICP-MS standards, allow for increased efficiency and accuracy of the analysis necessary to comply with the new regulations.

Use these standards can be used as a calibration or check standard to verify Oral Daily Dose PDE, Parenteral Component Limit or Parenteral Daily Dose PDE as well as Inhalation Component Limit or Daily Dose.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component standard contains 7 components.

• USP <232> Revision 40, Parenteral Mix 3 elemental impurities standard is a 125 mL bottle which contains 1,500 mg/kg (1,500 ppm) Molybdenum; 1,100 mg/kg (1,100 ppm) Chromium; 700 mg/kg (700 ppm) Barium; 600 mg/kg (600 ppm) Tin; 300 mg/kg (300 ppm) Copper; 250 mg/kg (250 ppm) Lithium; and 90 mg/kg (90 ppm) Antimony in 5% HNO3/tr. Tartaric Acid/tr. HF.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Matrix5% HNO3/tr. Tartaric Acid/tr. HF
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquids, toxic, n.o.s (Nitric acid solution, hydrofluoric acid); Class 8, 6.1, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion, Acute toxicity
  • UN NumberUN2922
  • Shelf Life (months)12
  • DescriptionUSP <232> Revision 40, Parenteral Mix 3 Elemental Impurities in 5% HNO3/tr. Tartaric Acid/tr. HF; 125 mL
Documents & Certificates
Reviews
Q&A