Spex THM-X Trihalomethanes Mix for US EPA Method 502.2, 200 µg/mL; 1 mL

Representative image only.

Spex THM-X Trihalomethanes Mix for US EPA Method 502.2, 200 µg/mL; 1 mL

Spex | Item: 0455627

Analytical standards for drinking water, wastewater, and solid waste

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 502.2
  • 200 ppm trihalomethanes mix in methanol-P&T matrix
  • 1 mL ampule

More About this Item

US EPA Method 502.2 is a general analytical method for the monitoring of halogenated, purgeable VOCs in drinking water and raw water sources by GC megabore capillary column techniques and Hall and Photoionization Detectors (PID).

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 4 components.

• Trihalomethanes Mix is a 1 mL ampule which contains 200 µg/mL (200 ppm) of Bromodichloromethane, Bromoform, Chloroform, and Dibromochloromethane.

Hazardous material shipping fees may apply.

Specifications
  • Concentration200 µg/mL (200 ppm)
  • MatrixMethanol-P&T
  • Element/IonBromodichloromethane, Bromoform, Chloroform, Dibromochloromethane
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Methanol; Class 3, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • UN NumberUN1230
  • CAS Number75-27-4, 75-25-2, 67-66-3, 124-48-1
  • Shelf Life (months)12
  • DescriptionTrihalomethanes Mix, 200 µg/mL in Methanol-P&T; 1 mL
Documents & Certificates
Reviews
Q&A