Spex TDS-2-500 Conductivity Standard, TDS as KCl, 1,000 µmhos; 500 mL

Representative image only.

Spex TDS-2-500 Conductivity Standard, TDS as KCl, 1,000 µmhos; 500 mL

Spex | Item: 0456984

Ensure accuracy by calibrating conductivity meters

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • ISO 17034 certified
  • 1,000 µmhos conductivity standard
  • 500 mL bottle

More About this Item

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Concentration1,000 µmhos
  • MatrixH2O
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)500
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • UN NumberNot regulated
  • Shelf Life (months)6
  • DescriptionConductivity Standard, Total Dissolved Solids as Potassium Chloride, 1,000 µmhos in H2O; 500 mL
Documents & Certificates
Reviews
Q&A