Spex TCLP-VX TCLP Volatiles Spike Standard, 2,000 µg/mL in Methanol-P&T; 1 mL

Representative image only.

Spex TCLP-VX TCLP Volatiles Spike Standard, 2,000 µg/mL in Methanol-P&T; 1 mL

Spex | Item: 0455687

Volatile organic standards for the monitoring of hazardous waste

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For TCLP analysis
  • 2,000 ppm TCLP volatiles spike standard in methanol-P&T matrix
  • 1 mL ampule

More About this Item

Toxicity Characteristic Leaching Procedure (TCLP) is a soil sample extraction method for chemical analysis employed as an analytical method to simulate leaching through a landfill.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-component solution contains 11 components.

• TCLP Volatiles Spike Standard is a 1 mL ampule which contains 2,000 µg/mL (2,000 ppm) of Benzene, 2-Butanone, Carbon tetrachloride, Chlorobenzene, Chloroform, 1,4-Dichlorobenzene, 1,2-Dichloroethane, 1,1-Dichloroethene, Tetrachloroethene, Trichloroethene, and Vinyl chloride.

Hazardous material shipping fees may apply.

Specifications
  • Concentration2,000 µg/mL (2,000 ppm)
  • MatrixMethanol-P&T
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Methanol; Class 3, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • UN NumberUN1230
  • CAS Number71-43-2, 78-93-3, 56-23-5, 108-90-7, 67-66-3, 106-46-7, 107-06-2, 75-35-4, 127-18-4, 79-01-6, 75-01-4
  • Shelf Life (months)12
  • DescriptionTCLP Volatiles Spike Standard, 2,000 µg/mL in Methanol-P&T; 1 mL
Documents & Certificates
Reviews
Q&A