Spex TCLP-BNA TCLP Base/Neutral/Acid Extractable Spike Solution, 2,000 µg/mL; 1 mL

Representative image only.

Spex TCLP-BNA TCLP Base/Neutral/Acid Extractable Spike Solution, 2,000 µg/mL; 1 mL

Spex | Item: 0455800

Analytical standards for drinking water, wastewater, and solid waste

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 1311 using TCLP analysis
  • 2,000 ppm TCLP base/neutral/acid extractable spike solution in methylene chloride matrix
  • 1 mL ampule

More About this Item

US EPA Method 1311-TCLP Series Methods - EPA methods 1311 and 1312 analytical methods for the monitoring of a wide range of compounds in various samples using the Toxicity Characteristic Leaching Procedure (TCLP).

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-component solution contains 13 components.

• TCLP Base/Neutral/Acid Extractable Spike Solution is a 1 mL ampule which contains 2,000 µg/mL (2,000 ppm) of 1,4-Dichlorobenzene, 2,4-Dinitrotoluen, Hexachlorobenzene, Hexachlorobutadiene, Hexachloroethane, 2-Methylphenol, 3-Methylphenol, 4-Methylphenol, Nitrobenzene, Pentachlorophenol, Pyridine, 2,4,5-Trichlorophenol, and 2,4,6-Trichlorophenol.

Hazardous material shipping fees may apply.

Specifications
  • Concentration2,000 µg/mL (2,000 ppm)
  • MatrixMethylene Chloride
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Dichloromethane; Class 6.1, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Irritant, Health Hazard
  • UN NumberUN1593
  • CAS Number106-46-7, 121-14-2, 118-74-1, 87-68-3, 67-72-1, 95-48-7, 108-39-4, 106-44-5, 98-95-3, 87-86-5, 110-86-1, 95-95-4, 88-06-2
  • Shelf Life (months)36
  • DescriptionTCLP Base/Neutral/Acid Extractable Spike Solution, 2,000 µg/mL in Methylene Chloride; 1 mL
Documents & Certificates
Reviews
Q&A