Spex SPXPR-3-PK Pesticide Mix 3 (15 Components), 100 µg/mL in LC/MS Acetonitrile; 1 mL; 4/pk

Representative image only.

Spex SPXPR-3-PK Pesticide Mix 3 (15 Components), 100 µg/mL in LC/MS Acetonitrile; 1 mL; 4/pk

Spex | Item: 0500948

Save time and money with premixed pesticide and multicompound CRMs

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 100 ppm pesticide mix 3 in LC/MS acetonitrile matrix
  • Four 1 mL ampules

More About this Item

Chemical pesticides have become an integral part of the agricultural toolbox, offering protection to crops from destructive pests. However, an unfortunate side effect of their use is the potential leaching of these, oftentimes, harmful chemicals into the environment leading to their eventual presence in the human food chain. As a result, pesticide residue analysis has become a critical testing process for many different types of laboratories.

Unfortunately, pesticide residue testing is a long, expensive and complicated process, covering hundreds of different compounds. As the leader in GC, GC/MS, HPLC, and LC/MS pesticide CRMs, Spex CertiPrep is happy to assist you with all of your pesticide CRM needs.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-compound standard contains 15 compounds.

• Pesticide Mix 3 is a 1 mL ampule which contains 100 µg/mL (100 ppm) of Carbaryl, Dimethomorph, Etofenprox, Etoxazole, Flonicamid, Methamidophos, Monocrotophos, Myclobutanil (Systhane), Phenthoate, Phorate, Pirimiphos-methyl, Profenofos, Propargite (Omite), Spirodiclofen, and Thiamethoxam.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethylene Chloride
  • ChemistryOrganics;Standards
  • Element/Ionn-Decane
  • FormLiquid
  • Volume (mL)1
  • CAS Number124-18-5
  • UN NumberUN1230
  • Shelf Life (months)36
  • Descriptionn-Decane Single-Component Organic Standard, 1,000 µg/mL (1,000 ppm), CAS #124-18-5 in Methylene Chloride, 1 mL; 4/PK
Documents & Certificates
Reviews
Q&A