Spex SPXMT-KIT Minerals Testing Kit, 1,000 µg/mL in 2% HNO3; 30 mL

Representative image only.

Spex SPXMT-KIT Minerals Testing Kit, 1,000 µg/mL in 2% HNO3; 30 mL

Spex | Item: 0456926

Routinely analyzed mineral standards in a convenient kit

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • Contains five minerals standards and one blank
  • Six 30 mL bottles

More About this Item

Minerals testing kit comes with six 30 mL standards plus a nitiric acid blank for easy dilution. Conveniently packaged in a sturdy, heavy-duty carton, these kits are perfect to store on a lab bench or in a cabinet. The 30 mL standards ship non-hazardous, saving money on shipping costs. The smaller volume also allows for less hazardous waste should the standard expire before its contents are used.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This minerals standard kit contains six standards.

• Standard 1 is a 30 mL bottle of 1,000 µg/mL (1,000 ppm) Calcium in 2% HNO3. This is available individually as part number 04502-70.
• Standard 2 is a 30 mL bottle of 1,000 µg/mL (1,000 ppm) Iron in 2% HNO3. This is available individually as part number 04502-77.
• Standard 3 is a 30 mL bottle of 1,000 µg/mL (1,000 ppm) Magnesium in 2% HNO3. This is available individually as part number 04502-80.
• Standard 4 is a 30 mL bottle of 1,000 µg/mL (1,000 ppm) Potassium in 2% HNO3. This is available individually as part number 04502-86.
• Standard 5 is a 30 mL bottle of 1,000 µg/mL (1,000 ppm) Sodium in 10% HNO3. This is available individually as part number 04502-91.
• Standard 6 is a 30 mL bottle of nitric acid blank in 2% HNO3. This is available individually as part number 04567-54.

Specifications
  • ConcentrationMultiple
  • Matrix2% HNO3
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)30
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolIrritant
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionMinerals Testing Kit, 1,000 µg/mL in 2% HNO3; 30 mL
Documents & Certificates
Reviews
Q&A