Spex SPXHM-KIT Heavy Metals Testing Kit, 1,000 µg/mL; 30 mL

Representative image only.

Spex SPXHM-KIT Heavy Metals Testing Kit, 1,000 µg/mL; 30 mL

Spex | Item: 0456925

Routinely analyzed heavy metal standards in a convenient kit

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • Contains five heavy metals standards and one blank
  • Six 30 mL bottles

More About this Item

Heavy metals testing kit comes with six 30 mL standards plus a nitiric acid blank for easy dilution. Conveniently packaged in a sturdy, heavy-duty carton, these kits are perfect to store on a lab bench or in a cabinet. The 30 mL standards ship non-hazardous, saving money on shipping costs. The smaller volume also allows for less hazardous waste should the standard expire before its contents are used.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This heavy metals standard kit contains six standards.

• Standard 1 is a 30 mL bottle of 1,000 µg/mL (1,000 ppm) Arsenic in 2% HNO3. This is available individually as part number 04502-64.
• Standard 2 is a 30 mL bottle of 1,000 µg/mL (1,000 ppm) Cadmium in 2% HNO3. This is available individually as part number 04506-69.
• Standard 3 is a 30 mL bottle of 1,000 µg/mL (1,000 ppm) Chromium in 2% HNO3. This is available individually as part number 04502-71.
• Standard 4 is a 30 mL bottle of 1,000 µg/mL (1,000 ppm) Lead in 2% HNO3. This is available individually as part number 04502-78.
• Standard 5 is a 30 mL bottle of 1,000 µg/mL (1,000 ppm) Mercury in 10% HNO3. This is available individually as part number 04502-83.
• Standard 6 is a 30 mL bottle of nitric acid blank in 2% HNO3. This is available individually as part number 04567-54.

Specifications
  • ConcentrationMultiple
  • MatrixMultiple
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)30
  • DOT InformationHazardous; Nitric acid solution; Class 8, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion, Health Hazard
  • UN NumberUN2031
  • Shelf Life (months)12
  • DescriptionHeavy Metals Testing Kit, 1,000 µg/mL; 30 mL
Documents & Certificates
Reviews
Q&A