Spex SPIKE-1 ICP Spike Sample 1 in 5% HNO3/tr. Tartaric Acid/tr. HF; 125 mL

Representative image only.

Spex SPIKE-1 ICP Spike Sample 1 in 5% HNO3/tr. Tartaric Acid/tr. HF; 125 mL

Spex | Item: 0456916

CLP Standards for ICP & ICP-MS instrumentation

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Provides all of the analytes required for the ICP-AES and the AA spike
  • Use in conjunction with the statement of work
  • Shows actual value of the solution, not a calculated number
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • Spike sample 1 in 5% HNO3/tr. tartaric acid/tr. HF matrix
  • 125 mL bottle

More About this Item

Our Contract Laboratory Program (CLP) standards allow you to Calibrate with Confidence®. The standards are to be used in conjunction with the Statement of Work for Inorganic Analysis; Multi-Media/Multi-Concentration Document Number ILM 05.3/ISM 01.2.

The final ICP check, performed in our own laboratories, is your stamp of assurance. We calibrate our instruments with traceable reference materials and show you the actual found value of the solution you receive, not just an ideal, calculated number as so many other standards manufacturers do. The combination of elements, concentrations and matrices listed have been diagnosed by Spex CertiPrep® for convenience of use and stability.

US EPA SOW ILM 05.3/ISM 01.2 gives specific procedures for the methods of analysis, target elements, and concentration levels. Standards are specified not only by the elements present and their relative concentrations, but also the order and frequency of running standards, blanks and samples. Details of these specifications may be found in the US EPA SOW ILM 05.3/ISM 01.2 in the following sections:

• Exhibit C, Inorganic Target Analyte List (TAL)
• Exhibit D, Analytical Methods
• Exhibit E, QA/QC Requirements

CLP ILM 02.0 Standards for ICP
ICP Spike Sample 1 standard provides all of the analytes required for the ICP-AES and the AA spike. Add 1 mL of ICP Spike Sample 1 Standard to aqueous samples and 2 mL to solid samples prior to digestion.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-element standard contains 18 components.

• ICP spike sample 1 is a 125 mL bottle which contains 200 µg/mL (200 ppm) of Aluminum, Arsenic, Barium, Selenium, Thallium; 100 µg/mL (100 ppm) of Iron; 50 µg/mL (50 ppm) of Antimony, Cobalt, Lead, Manganese, Nickel, Vanadium, Zinc; 20 µg/mL (20 ppm) of Copper; 20 µg/mL (20 ppm) of Chromium; and 5 µg/mL (5 ppm) of Beryllium, Cadmium, Silver.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Matrix5% HNO3/tr. Tartaric Acid/tr. HF
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionICP Spike Sample 1 in 5% HNO3/tr. Tartaric Acid/tr. HF; 125 mL
Documents & Certificates
Reviews
Q&A