Spex SPEC-AS3M Assurance® Grade Arsenic +3, 1,000 µg/mL in 2% HCl; 30 mL

Representative image only.

Spex SPEC-AS3M Assurance® Grade Arsenic +3, 1,000 µg/mL in 2% HCl; 30 mL

Spex | Item: 0450458

Speciation standards for analyzing species in a sample

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • 1,000 ppm arsenic +3 speciation standard in 2% HCl matrix
  • 30 mL bottle

More About this Item

Speciation analysis has become common in many fields, including environmental, food and pharmaceutical testing labs. To analyze species within a sample requires Certified Reference Materials (CRMs) for sample verification and method validation. Many speciation standards are available in today’s market, however, most of them are not certified or analyzed with a state-of-the-art ICP, ICP-MS or LC-ICP-MS. SPEX CertiPrep offers a wide variety of speciation standards, certified to the strictest ISO 17034 guidelines, and tested on our own LC-ICP-MS.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Purity GradeAssurance® Grade
  • Concentration1,000 µg/mL (1,000 ppm)
  • Matrix2% HCl
  • Element/IonArsenic +3 (As)
  • FormLiquid
  • Volume (mL)30
  • DOT InformationHazardous; Hydrochloric acid; Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolHealth Hazard
  • UN NumberUN1789
  • Shelf Life (months)12
  • DescriptionAssurance® Grade Arsenic +3 Speciation Standard, 1,000 µg/mL in 2% HCl; 30 mL
Documents & Certificates
Reviews
Q&A