Spex SDFS-SET Sullfur Oil Standard Set in #2 Diesel Fuel, 8 Bottles

Representative image only.

Spex SDFS-SET Sullfur Oil Standard Set in #2 Diesel Fuel, 8 Bottles

Spex | Item: 0457045

For use in diesel fuel analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Certified by DQS to ISO 9001:2015
  • Sulfur oil standard set in #2 diesel fuel contains 8 standards

More About this Item

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This sulfur oil standard set contains 8 solutions.

• Solution 1 is a 100 mL bottle which contains 100 µg/g (100 ppm) of Sulfur. This is available individually as part number 04570-54.
• Solution 2 is a 100 mL bottle which contains 50 µg/g (50 ppm) of Sulfur. This is available individually as part number 04570-52.
• Solution 3 is a 100 mL bottle which contains 25 µg/g (25 ppm) of Sulfur. This is available individually as part number 04570-51.
• Solution 4 is a 100 mL bottle which contains 20 µg/g (20 ppm) of Sulfur. This is available individually as part number 04570-50.
• Solution 5 is a 100 mL bottle which contains 15 µg/g (15 ppm) of Sulfur. This is available individually as part number 04570-49.
• Solution 6 is a 100 mL bottle which contains 10 µg/g (10 ppm) of Sulfur. This is available individually as part number 04570-48.
• Solution 7 is a 100 mL bottle which contains 5 µg/g (5 ppm) of Sulfur. This is available individually as part number 04570-47.
• Solution 8 is a 100 mL bottle Sulfur blank. This is available individually as part number 04570-46.

Specifications
  • ConcentrationMultiple
  • Matrix#2 Diesel Fuel
  • Element/IonEight bottle set
  • FormLiquid
  • Volume (mL)100
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • Hazard SymbolIrritant, Health Hazard
  • UN NumberNot regulated
  • Shelf Life (months)12
  • DescriptionSullfur Oil Standard Set for Diesel Fuel Analysis in #2 Diesel Fuel, 8 Bottles
Documents & Certificates
Reviews
Q&A