Spex SDFS-1000-Y Sulfur Oil Standard, 1,000 µg/g in #2 Diesel Fuel; 100 mL

Representative image only.

Spex SDFS-1000-Y Sulfur Oil Standard, 1,000 µg/g in #2 Diesel Fuel; 100 mL

Spex | Item: 0457060

For use in diesel fuel analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Certified by DQS to ISO 9001:2015
  • 1,000 ppm sulfur oil standard for diesel fuel analysis in #2 diesel fuel matrix
  • 100 mL bottle

More About this Item

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Concentration1,000 µg/g (1,000 ppm)
  • Matrix#2 Diesel Fuel
  • Element/IonSulfur
  • FormLiquid
  • Volume (mL)100
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • Hazard SymbolIrritant, Health Hazard
  • UN NumberNot regulated
  • Shelf Life (months)12
  • DescriptionSulfur Oil Standard for Diesel Fuel Analysis, 1,000 µg/g in #2 Diesel Fuel; 100 mL
Documents & Certificates
Reviews
Q&A