Spex S-930-2K Chloroprene Standard for US EPA Method 8260B, 2,000 µg/mL; 1 mL

Representative image only.

Spex S-930-2K Chloroprene Standard for US EPA Method 8260B, 2,000 µg/mL; 1 mL

Spex | Item: 0455675

Analytical standards for monitoring VOCs by GC/MS

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 8260B
  • 2,000 ppm chloroprene standard in methanol-P&T matrix
  • 1 mL ampule

More About this Item

US EPA Method 8260 is a general analytical method for the monitoring of VOCs by GC/MS.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration2,000 µg/mL (2,000 ppm)
  • MatrixMethanol-P&T
  • Element/IonChloroprene (2-chloro-1,3-butadiene)
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Flammable liquid, toxic, n.o.s. (Methanol); Class 3, 6.1, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • UN NumberUN1992
  • CAS Number126-99-8
  • Shelf Life (months)18
  • DescriptionChloroprene Standard for US EPA Method 8260B, 2,000 µg/mL in Methanol-P&T; 1 mL
Documents & Certificates
Reviews
Q&A