Spex S-4871 Tetraethyl Lead, 1,000 µg/m in Methanol; 1 mL

Representative image only.

Spex S-4871 Tetraethyl Lead, 1,000 µg/m in Methanol; 1 mL

Spex | Item: 0455472

Single-component organic standards ensure consistent results

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm tetraethyl lead in methanol matrix

More About this Item

Single-component standards offer high-quality, consistent results while saving preparation time. Tetraethyl lead standard is supplied in methanol. Additional solvents include acetone, methylene chloride, hexane, benzene, acetonitrile, THF, and water. Please inquire about the matrix for a particular item when ordering, if the matrix is important to your application, a substitution may be possible.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethanol
  • Element/IonTetraethyl lead
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Methanol; Class 3, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • CAS Number78-00-2
  • UN NumberUN1230
  • Shelf Life (months)12
  • DescriptionTetraethyl Lead, 1,000 µg/m in Methanol; 1 mL
Documents & Certificates
Reviews
Q&A