Spex S-3192 Propionic Acid Organic Standard, 1,000 µg/mL in DI Water; 1 mL

Representative image only.

Spex S-3192 Propionic Acid Organic Standard, 1,000 µg/mL in DI Water; 1 mL

Spex | Item: 0455356

Single-component organic standards ensure consistent results

£45.00GBP / Each
£54.00GBP (incl VAT)

£45.00GBP / Each
£54.00GBP (incl VAT)
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Certified by DQS to ISO 9001:2015
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm propionic acid in DI water matrix
  • 1 mL ampule

More About this Item

Single-component standards offer high-quality, consistent results while saving preparation time. Propionic acid standard is supplied in DI water. Additional solvents include methylene chloride, methanol, benzene, hexane, acetonitrile, THF, and acetone. Please inquire about the matrix for a particular item when ordering, if the matrix is important to your application, a substitution may be possible.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixDI Water
  • Element/IonPropionic acid
  • FormLiquid
  • Volume (mL)1
  • DOT InformationNot Regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • CAS Number79-09-4
  • UN NumberNot Regulated
  • Shelf Life (months)36
  • DescriptionPropionic Acid Organic Standard, 1,000 µg/mL in DI Water; 1 mL
Documents & Certificates
Reviews
Q&A