Spex S-2635 Mirex, 1,000 µg/mL in Hexane:Toluene (50:50); 1 mL

Representative image only.

Spex S-2635 Mirex, 1,000 µg/mL in Hexane:Toluene (50:50); 1 mL

Spex | Item: 0455140

Single-component organic standards ensure consistent results

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm mirex in hexane:toluene (50:50) matrix
  • 1 mL ampule

More About this Item

Single-component standards offer high-quality, consistent results while saving preparation time. Select mirex standard is supplied in acetonitrile or hexane:toluene (50:50). Additional solvents include acetone, methanol, methylene chloride, benzene, hexane, acetonitrile, THF, and water. Please inquire about the matrix for a particular item when ordering, if the matrix is important to your application, a substitution may be possible.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixHexane:Toluene (50:50)
  • Element/IonMirex
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Flammable liquids, n.o.s. (Hexanes, Toluene); Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Irritant, Health Hazard
  • UN NumberUN1993
  • CAS Number2385-85-5
  • Shelf Life (months)36
  • DescriptionMirex, 1,000 µg/mL in Hexane:Toluene (50:50); 1 mL
Documents & Certificates
Reviews
Q&A