Spex S-2455 Methyl Tertiary-butyl Ether, 1,000 µg/mL in Methanol-P&T; 1 mL

Representative image only.

Spex S-2455 Methyl Tertiary-butyl Ether, 1,000 µg/mL in Methanol-P&T; 1 mL

Spex | Item: 0455082

Single-component organic standards ensure consistent results

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Certified by DQS to ISO 9001:2015
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm methyl tertiary-butyl ether in methanol-P&T matrix
  • 1 mL ampule

More About this Item

Single-component standards offer high-quality, consistent results while saving preparation time. Methyl tertiary-butyl ether standard is supplied in methanol-P&T. Additional solvents include methylene chloride, acetone, benzene, hexane, acetonitrile, THF, and water. Please inquire about the matrix for a particular item when ordering, if the matrix is important to your application, a substitution may be possible.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethanol-P&T
  • Element/IonMethyl tertiary-butyl ether
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Methanol; Class 3, 6.1, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • CAS Number1634-04-4
  • UN NumberUN1230
  • Shelf Life (months)36
  • Boiling Point C (° C)55
  • Boiling Point F (° F)131
  • Polarity Index2.5
  • Solubility in Water (% wt/wt)4.8
  • Specific Gravity0.74
  • UV Cutoff (nm)210
  • DescriptionMethyl Tertiary-butyl Ether Organic Standard, 1,000 µg/mL in Methanol-P&T; 1 mL
Documents & Certificates
Reviews
Q&A