Spex S-2332-PK Isovaleric Acid Organic Standard, 1,000 µg/mL in DI Water; 1 mL; 4/pk

Representative image only.

Spex S-2332-PK Isovaleric Acid Organic Standard, 1,000 µg/mL in DI Water; 1 mL; 4/pk

Spex | Item: 0500784

Single-component organic standards ensure consistent results

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm isovaleric acid single-component organic standard in DI water matrix
  • Four 1 mL ampules

More About this Item

Single-component standards offer high-quality, consistent results while saving preparation time. Isovaleric acid standard is supplied in DI water. Additional solvents include methanol, methylene chloride, acetone, benzene, hexane, acetonitrile, and THF. Please inquire about the matrix for a particular item when ordering, if the matrix is important to your application, a substitution may be possible.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • MatrixHPLC Acetonitrile
  • ChemistryStandards;Organics
  • Element/IonSpirotetramat
  • FormLiquid
  • Volume (mL)1
  • Packaging4 x 1 mL
  • DOT InformationHazardous; Acetonitrile; Class 3, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Irritant
  • CAS Number203313-25-1
  • UN NumberUN1648
  • Shelf Life (months)24
  • DescriptionSpirotetramat Organic Standard, 1,000 µg/mL in HPLC Acetonitrile; 1 mL
Documents & Certificates
Reviews
Q&A