Supplier Certificate of Analysis (CoA) is included with product shipment.
Key Features
Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
1,000 ppm n-hexane in methanol-P&T matrix
Molecular formula C6H14
1 mL ampule
More About this Item
The EPA regulates the monitoring and testing of underground storage tanks (USTs), that were or are used to store petroleum or other hazardous substances, which could impact the environment and human health. Currently, there are over half a million registered underground storage tanks in the US alone. Many tanks installed prior to the mid-80s were composed of steel, which can corrode over time allowing for leakage into the surrounding groundwater and soil.
The Hazardous and Solid Waste Amendments to the Resource Conservation and Recovery Act (RCRA), passed in 1984, required the US Environmental Protection Agency (EPA) to develop regulations for the underground storage of motor fuels. The EPA requires owners and operators of USTs to maintain and clean up sites damaged by petroleum contamination.
Spex CertiPrep® provides standards designed to meet the testing requirements set up by the EPA and many individual state regulations for leaking underground storage tanks and leaking underground fuel tanks.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Hazardous material shipping fees may apply.
Specifications
Concentration1,000 µg/mL (1,000 ppm)
MatrixMethanol-P&T
Element/Ionn-Hexane
FormLiquid
Volume (mL)1
DOT InformationHazardous; Methanol; Class 3, Packing Group II
Storage SymbolAmbient
Hazard SymbolFlammable, Acute Toxicity, Health Hazard
CAS Number110-54-3
UN NumberUN1230
Shelf Life (months)36
Boiling Point C (° C)69
Boiling Point F (° F)156.2
Melting Point C (° C)-95
Melting Point F (° F)-139
Polarity Index0
Solubility in Water (% wt/wt)1E-05
Specific Gravity0.659
UV Cutoff (nm)200
Descriptionn-Hexane, 1,000 µg/mL in Methanol-P&T, Formula C6H14; 1 mL