Spex S-1890 Ethanolamine Organic Standard, 1,000 µg/mL in Ethanol; 1 mL

Representative image only.

Spex S-1890 Ethanolamine Organic Standard, 1,000 µg/mL in Ethanol; 1 mL

Spex | Item: 0454787

Single-component organic standards ensure consistent results

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm ethanolamine single-component organic standard in ethanol matrix
  • 1 mL ampule

More About this Item

Single-component standards offer high-quality, consistent results while saving preparation time. Ethanolamine standard is supplied in ethanol. Additional solvents include methanol, methylene chloride, acetone, benzene, hexane, acetonitrile, THF, and water. Please inquire about the matrix for a particular item when ordering, if the matrix is important to your application, a substitution may be possible.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixEthanol
  • Element/IonEthanolamine
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Ethanol; Class 3, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Health Hazard
  • CAS Number141-43-5
  • UN NumberUN1170
  • Shelf Life (months)12
  • DescriptionEthanolamine Organic Standard, 1,000 µg/mL in Ethanol; 1 mL
Documents & Certificates
Reviews
Q&A