Supplier Certificate of Analysis (CoA) is included with product shipment.
Key Features
Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
1,000 ppm (-)-delta9-THC DEA controlled substance in methanol matrix
More About this Item
While the legalization of cannabis for both medicinal and recreational purposes has been gaining speed, legislation and regulation has not necessarily kept pace. Out of a drive for self-regulation and significant consumer safety concerns, many producers and manufacturers are turning to testing labs in order to ensure that their products are of high quality and free of chemical contaminants.
Spex CertiPrep® offers ISO/IEC 17025 and ISO 17034 Certified Reference Materials (CRMs) for all of the common contaminants such as pesticide residues, residual solvents and heavy metals, as well as qualitative analysis CRMs, such as terpenes. As the industry demands change and regulations are put into place, we continually update our product offering.
Designed for state specific pesticide regulations: • OAR 333-008-11 • HB 3460 • AOAC 2007-01 • EN 15662
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Hazardous material shipping fees may apply.
NOTE: This product ships overnight.
Specifications
Concentration1,000 µg/mL (1,000 ppm)
MatrixMethanol
Element/Ion(-)-delta9-THC
FormLiquid
Volume (mL)1
DOT InformationHazardous; Methanol; Class 3, Packing Group II
Storage SymbolFreezer
Hazard SymbolFlammable, Acute Toxicity, Health Hazard
UN NumberUN1230
CAS Number1972-08-3
Shelf Life (months)12
Description(-)-delta9-THC DEA Controlled Substance, 1,000 µg/mL in Methanol; 1 mL