Spex S-10248 Cannabichromene (CBC) DEA Controlled Substance, 1,000 µg/mL; 1 mL

Representative image only.

Spex S-10248 Cannabichromene (CBC) DEA Controlled Substance, 1,000 µg/mL; 1 mL

Spex | Item: 0455962

Analytical standards for medicinal and recreational cannabis testing helps ensure high-quality products

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 1,000 ppm cannabichromene (CBC) DEA controlled substance in methanol matrix

More About this Item

While the legalization of cannabis for both medicinal and recreational purposes has been gaining speed, legislation and regulation has not necessarily kept pace. Out of a drive for self-regulation and significant consumer safety concerns, many producers and manufacturers are turning to testing labs in order to ensure that their products are of high quality and free of chemical contaminants.

Spex CertiPrep® offers ISO/IEC 17025 and ISO 17034 Certified Reference Materials (CRMs) for all of the common contaminants such as pesticide residues, residual solvents and heavy metals, as well as qualitative analysis CRMs, such as terpenes. As the industry demands change and regulations are put into place, we continually update our product offering.

Designed for state specific pesticide regulations:
• OAR 333-008-11
• HB 3460
• AOAC 2007-01
• EN 15662

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

NOTE: This product ships overnight.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethanol
  • Element/IonCannabichromene (CBC)
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Methanol; Class 3, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Acute Toxicity, Health Hazard
  • UN NumberUN1230
  • CAS Number20675-51-8
  • Shelf Life (months)24
  • DescriptionCannabichromene (CBC) DEA Controlled Substance, 1,000 µg/mL in Methanol; 1 mL
Documents & Certificates
Reviews
Q&A