Spex S-030W-20K Motor Oil, SAW 30W Petrochemical Fuel Standard, 20,000 µg/mL; 1mL

Representative image only.

Spex S-030W-20K Motor Oil, SAW 30W Petrochemical Fuel Standard, 20,000 µg/mL; 1mL

Spex | Item: 0456115

Test for groundwater and soil contamination caused by underground storage tanks

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • 20,000 ppm motor oil, SAW 30W petrochemical fuel standard in methylene chloride matrix

More About this Item

The EPA regulates the monitoring and testing of underground storage tanks (USTs), that were or are used to store petroleum or other hazardous substances, which could impact the environment and human health. Currently, there are over half a million registered underground storage tanks in the US alone. Many tanks installed prior to the mid-80s were composed of steel, which can corrode over time allowing for leakage into the surrounding groundwater and soil.

The Hazardous and Solid Waste Amendments to the Resource Conservation and Recovery Act (RCRA), passed in 1984, required the US Environmental Protection Agency (EPA) to develop regulations for the underground storage of motor fuels. The EPA requires owners and operators of USTs to maintain and clean up sites damaged by petroleum contamination.

Spex CertiPrep® provides standards designed to meet the testing requirements set up by the EPA and many individual state regulations for leaking underground storage tanks and leaking underground fuel tanks.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

Specifications
  • Concentration20,000 µg/mL (20,000 ppm)
  • MatrixMethylene Chloride
  • Element/IonMotor Oil, SAW 30W
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Dichloromethane; Class 6.1, Packing Group III
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Irritant, Health Hazard
  • CAS NumberN/A
  • UN NumberUN1593
  • Shelf Life (months)36
  • DescriptionMotor Oil, SAW 30W Petrochemical Fuel Standard, 20,000 µg/mL in Methylene Chloride; 1 mL
Documents & Certificates
Reviews
Q&A