Spex RSCN9-SET Cyanide (CN-) Ion Selective Electrode Standard Set, 2 Solution Set

Representative image only.

Spex RSCN9-SET Cyanide (CN-) Ion Selective Electrode Standard Set, 2 Solution Set

Spex | Item: 0456941

Single-element standard for ion chromatography

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Ideal for calibrating and performing QA/QC checks on ion chromatography systems
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • Set of two solutions: sodium hydroxide buffer and simple cyanide

More About this Item

Ion chromatography (IC) is an analytical process for the separation of ions based on charge affinity. IC can be used for a variety of different kinds of charged analytes from single elements to large proteins. In order to ensure accurate analysis, quality standards which are traceable and stable are necessary. Spex CertiPrep® IC standards are of the highest quality available for the analytical laboratory.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This standard set contains 2 solutions.

• Solution 1 is a 500 mL bottle of 10 M sodium hydroxide ionic strength adjustment buffer. This is available individually as part number 04569-28.

• Solution 2 is a 125 mL bottle of 1,000 µg/mL (1,000 ppm) cyanide. This is available individually as part number 04501-58.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • MatrixMultiple
  • Element/IonCyanide and Sodium hydroxide
  • FormLiquid
  • DOT InformationHazardous; Sodium hydroxide solution; Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN1814
  • DescriptionSet of 2 Solutions for Cyanide
Documents & Certificates
Reviews
Q&A