Spex QC-SETB Assurance® Grade Quality Control Standard Set B for AA&ICP; 125mL

Representative image only.

Spex QC-SETB Assurance® Grade Quality Control Standard Set B for AA&ICP; 125mL

Spex | Item: 0456809

Multi-element standards for AA and ICP analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • Quality control standard set contain 2 solutions
  • QC Standard Set B contains QC Standard 21 (04568-00) and QC Standard 7 (04567-96)
  • Two 125 mL bottles

More About this Item

Quality Control Standards are used to check the standard curve, the procedure for inter-element correction and other spectral interferences. These standards are carried through the entire analytical operation of the method. If the determined concentration is not within ±5% of 1 µg/mL, the laboratory performance is unacceptable. The source of the problem should be identified and corrected before continuing the analysis.

Standards may be diluted in the same matrix as specified; however, caution must be exercised in the choice of the source for your diluents. Diluting the matrix may cause some standards to precipitate. Also, an impure or unknown diluent turns your standard into an unknown. We recommend using only SPEX CertiPrep Matrix Blanks when diluting your standards.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-element standard set contains 2 solutions.

• Solution 1 (QC standard 21) is a 125 mL bottle which contains 100 µg/mL (100 ppm) Antimony, Arsenic, Beryllium, Cadmium, Calcium, Chromium, Cobalt, Copper, Iron, Lead, Lithium, Magnesium, Manganese, Molybdenum, Nickel, Selenium, Strontium, Thallium, Titanium, Vanadium, Zinc in 5% HNO3/tr. Tartaric Acid/tr. HF. This is available individually as part number 04568-00.

• Solution 2 (QC standard 7) is a 125 mL bottle which contains 1,000 µg/mL (1,000 ppm) Potassium; 100 µg/mL (100 ppm) Aluminum, Barium, Boron, Silver, Sodium; and 50 µg/mL (50 ppm) Silicon in 5% HNO3/tr. F-. This is available individually as part number 04567-96.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Purity GradeAssurance® Grade
  • MatrixMultiple
  • Element/IonTwo bottle set: Solution 1 and Solution 3
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionAssurance® Grade Quality Control Standard Set B for AA & ICP; 125 mL
Documents & Certificates
Reviews
Q&A