Spex QC-7A Assurance® Grade Quality Control Standard 7A for AA & ICP in 5% HNO3/tr. HF; 125 mL

Representative image only.

Spex QC-7A Assurance® Grade Quality Control Standard 7A for AA & ICP in 5% HNO3/tr. HF; 125 mL

Spex | Item: 0456798

Multi-element standards for AA & ICP instrumentation

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Quality control standards help ensure accurate and reliable results
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • Quality control standard 7A in 5% HNO3/tr. F- matrix
  • 125 mL bottle

More About this Item

Use the Quality Control Standards to check the standard curve, the procedure for inter-element correction and other spectral interferences. These standards are carried through the entire analytical operation of the method. If the determined concentration is not within ± 5% of 1 µg/mL, the laboratory performance is unacceptable. The source of the problem should be identified and corrected before continuing the analysis.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-element standard contains 7 components.

• Quality control standard 7A is a 125 mL bottle which contains 1,000 µg/mL (1,000 ppm) of Potassium; 500 µg/mL (500 ppm) of Silicon; 100 µg/mL (100 ppm) of Aluminum, Barium, Boron, Sodium; and 50 µg/mL (50 ppm) of Silver.

Hazardous material shipping fees may apply.

Specifications
  • Purity GradeAssurance® Grade
  • ConcentrationMultiple
  • Matrix5% HNO3/tr. HF
  • Element/IonAluminum, Barium, Boron, Potassium, Silicon, Silver, and Sodium
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionAssurance® Grade Quality Control Standard 7A for AA & ICP in 5% HNO3/tr. HF; 125 mL
Documents & Certificates
Reviews
Q&A