Spex QC-24 Assurance® Grade Quality Control Standard 24 for AA & ICP; 125 mL

Representative image only.

Spex QC-24 Assurance® Grade Quality Control Standard 24 for AA & ICP; 125 mL

Spex | Item: 0456807

Multi-element quality control standard for AA and ICP instrumentation

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025/2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • 10 ppm quality control standard 24 for AA & ICP in 10% HNO3 matrix
  • 125 mL bottle

More About this Item

Quality Control Standards are used to check the standard curve, the procedure for inter-element correction and other spectral interferences. These standards are carried through the entire analytical operation of the method. If the determined concentration is not within ±5% of 1 µg/mL, the laboratory performance is unacceptable. The source of the problem should be identified and corrected before continuing the analysis.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-element standard contains 22 elements.

• Quality Control Standard 24 is a 125 mL bottle which contains 10 µg/mL (10 ppm) of Aluminum, Barium, Bismuth, Boron, Cadmium, Calcium, Chromium, Cobalt, Copper, Gallium, Indium, Iron, Lead, Lithium, Magnesium, Manganese, Nickel, Potassium, Silver, Sodium, Thallium, Zinc.

Hazardous material shipping fees may apply.

Specifications
  • Concentration10 µg/mL (10 ppm)
  • Purity GradeAssurance® Grade
  • Matrix10% HNO3
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Nitric acid solution; Class 8, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN2031
  • Shelf Life (months)12
  • DescriptionAssurance® Grade Quality Control Standard 24 for AA & ICP, 10 µg/mL in 10% HNO3; 125 mL
Documents & Certificates
Reviews
Q&A