Spex QC-21-250 Assurance® Grade Quality Control Standard 21, 100 µg/mL for AA & ICP in 5% HNO3 Tartaric Acid HF; 250 mL

Representative image only.

Spex QC-21-250 Assurance® Grade Quality Control Standard 21, 100 µg/mL for AA & ICP in 5% HNO3 Tartaric Acid HF; 250 mL

Spex | Item: 0456801

Mixed multi-element standard for AA & ICP

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Diluted in 5% HNO3/tr. Tartaric Acid/tr. HF
  • Use standard to check spectral interferences

More About this Item

Standards may be diluted in the same matrix as specified; however, caution must be exercised in the choice of the source for your diluents. Diluting the matrix may cause some standards to precipitate. Also, an impure or unknown diluent turns your standard into an unknown. We recommend using only Spex CertiPrep® Matrix Blanks when diluting your standards.

Quality Control Standards are used to check the standard curve, the procedure for inter-element correction and other spectral interferences. These standards are carried through the entire analytical operation of the method. If the determined concentration is not within +/- 5% of 1 µg/mL, the laboratory performance is unacceptable. The source of the problem should be identified and corrected before continuing the analysis.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

Specifications
  • Concentration100 µg/mL (100 ppm)
  • Purity GradeAssurance® Grade
  • Matrix5% HNO3/tr. Tartaric Acid/tr. HF
  • ClassAA/ICP
  • Element/IonAntimony, Arsenic, Beryllium, Cadmium, Calcium, Chromium, Cobalt, Copper, Iron, Lead, Lithium, Magnesium, Manganese, Molybdenum, Nickel, Selenium, Strontium, Thallium, Titanium, Vanadium, Zinc
  • FormLiquid
  • Volume (mL)250
  • DOT InformationCorrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution)
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN3264
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionQuality Control Standard 21, 100 µg/mL for AA & ICP in 5% HNO3 Tartaric Acid HF; 250 mL
Documents & Certificates
Reviews
Q&A