Spex PLTI9-2M Assurance® Grade Titanium, 1,000 µg/mL for AA and ICP in H2O/0.24% F-; 30 mL

Representative image only.

Spex PLTI9-2M Assurance® Grade Titanium, 1,000 µg/mL for AA and ICP in H2O/0.24% F-; 30 mL

Spex | Item: 0450523

Single-element standards for AA and ICP analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Made with acid and ASTM Type I Water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • 1,000 ppm titanium in H2O/0.24% F- matrix
  • 30 mL bottle

More About this Item

Assurance® grade certified reference materials (CRMs) are designed for atomic absorption (AA) and inductively coupled plasma (ICP) analysis. Custom standards can be manufactured upon request.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • Purity GradeAssurance® Grade
  • MatrixH2O/0.24% F-
  • Element/IonTitanium (Ti)
  • FormLiquid
  • Volume (mL)30
  • DOT InformationNon-Hazardous; Not regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • UN NumberNot regulated
  • Shelf Life (months)12
  • DescriptionAssurance® Grade Titanium, 1,000 µg/mL for AA and ICP in H2O/0.24% F-; 30 mL
Documents & Certificates
Reviews
Q&A