Spex PLTA9-2M Assurance® Grade Tantalum, 1,000 µg/mL in H2O/0.8% HF; 30 mL

Representative image only.

Spex PLTA9-2M Assurance® Grade Tantalum, 1,000 µg/mL in H2O/0.8% HF; 30 mL

Spex | Item: 0450516

Single-element standards for AA and ICP analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Made with acid and ASTM Type I Water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • 1,000 ppm tantalum in H2O/0.8% HF matrix
  • 30 mL bottle

More About this Item

Assurance® grade certified reference materials (CRMs) are designed for atomic absorption (AA) and inductively coupled plasma (ICP) analysis.

Can’t find the standards you are looking for? Spex CertiPrep can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • Purity GradeAssurance® Grade
  • MatrixH2O/0.8% HF
  • Element/IonTantalum (Ta)
  • FormLiquid
  • Volume (mL)30
  • DOT InformationHazardous; Corrosive liquids, toxic, n.o.s (Hydrofluoric acid); Class 8, 6.1, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolAcute toxicity, Irritant
  • UN NumberUN2922
  • Shelf Life (months)12
  • DescriptionAssurance® Grade Tantalum, 1,000 µg/mL for AA and ICP in H2O/0.8% HF; 30 mL
Documents & Certificates
Reviews
Q&A