Spex PLSI9-2X Assurance® Grade Silicon, 1,000 µg/mL (1,000 ppm) for AA and ICP in Water & 0.4% Fluoride; 500 mL

Representative image only.

Spex PLSI9-2X Assurance® Grade Silicon, 1,000 µg/mL (1,000 ppm) for AA and ICP in Water & 0.4% Fluoride; 500 mL

Spex | Item: 0450631

Single-element standards for AA and ICP analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Made with acid and ASTM Type I water
  • Inorganic compounds at 99.99% to 99.9999% purity
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • 1,000 ppm Silicone in water and 0.4% Fluoride matrix

More About this Item

Assurance® grade certified reference materials (CRMs) are designed for atomic absorption (AA) and inductively coupled plasma (ICP) analysis. Custom standards can be manufactured upon request.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Specifications
  • Concentration1,000 µg/mL in water & 0.4% F
  • Purity GradeAssurance® Grade
  • MatrixWater & 0.4% fluoride
  • ClassAA & ICP
  • Element/IonSilicon (Si)
  • FormLiquid
  • Volume (mL)500
  • Storage SymbolAmbient
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionAssurance® Grade Silicon, 1,000 µg/mL (1,000 ppm) for AA and ICP in Water & 0.4% Fluoride; 500 mL
Documents & Certificates
Reviews
Q&A