Spex PLPT3-2Y Assurance® Grade Platinum, 1,000 µg/mL, for AA and ICP in 10% HCl; 125 mL

Representative image only.

Spex PLPT3-2Y Assurance® Grade Platinum, 1,000 µg/mL, for AA and ICP in 10% HCl; 125 mL

Spex | Item: 0450372

Single-element standards for AA and ICP analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Made with acid and ASTM Type I water
  • Inorganic compounds at 99.99% to 99.9999% purity
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • 1,000 ppm Platinum in 10% HCl matrix

More About this Item

Assurance® grade certified reference materials (CRMs) are designed for atomic absorption (AA) and inductively coupled plasma (ICP) analysis. Custom standards can be manufactured upon request.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixHCl
  • Element/IonPlatinum
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHydrochloric acid; Class 8, Packing Group II
  • Hazard SymbolIrritant
  • UN NumberUN1789
  • Shelf Life (months)12
  • DescriptionAssurance® Grade Platinum, 1,000 µg/mL, for AA and ICP in 10% HCl; 125 mL
Documents & Certificates
Reviews
Q&A