Spex PLAL2-2T Assurance® Grade Aluminum, 1,000 µg/mL for AA and ICP in 2% HNO3; 250 mL

Representative image only.

Spex PLAL2-2T Assurance® Grade Aluminum, 1,000 µg/mL for AA and ICP in 2% HNO3; 250 mL

Spex | Item: 0450421

Single-element standards for AA and ICP analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Made with acid and ASTM Type I water
  • Inorganic compounds and metals at 99.99% to 99.9999% purity
  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016

More About this Item

Assurance® grade certified reference materials (CRMs) are designed for atomic absorption (AA) and inductively coupled plasma (ICP) analysis. They are packaged in 30 mL, 125 mL, 250 mL, and 500 mL bottles to minimize contamination.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

CorrosiveIrritant
Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • Purity GradeAssurance® Grade
  • Matrix2% HNO3
  • ClassAA/ICP
  • Element/IonAl
  • FormLiquid
  • Volume (mL)250
  • DOT InformationHazardous
  • Storage SymbolAmbient
  • Hazard SymbolWarning
  • UN NumberUN3264
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionAluminum, 1,000 µg/mL, for AA and ICP, 250 mL
Documents & Certificates
Reviews
Q&A