Spex PH-BUFF5-500 pH 5 Certified Buffer Standard, 5 SI Units in H2O, 500 mL

Representative image only.

Spex PH-BUFF5-500 pH 5 Certified Buffer Standard, 5 SI Units in H2O, 500 mL

Spex | Item: 0456975

Premium grade for calibration and quality control checks

Call For Price

Call For Price

Buy It Together

Current Item

Spex PH-BUFF5-500 pH 5 Certified Buffer Standard, 5 SI Units in H2O, 500 mL
Spex PH-BUFF5-500 pH 5 Certified Buffer Standard, 5 SI Units in H2O, 500 mL
0456975

Call For Price
+ =
Total Price 48.21 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Analyzed for certified value and homogeneity
  • Certified to ISO/IEC 17025:2005 and ISO 17034:2016

More About this Item

Spex CertiPrep® pH buffers are accurate and certified to ISO/IEC 17025:2005 and ISO 17034:2016 requirements. Made using high purity starting materials and ASTM Type I water, they have been thoroughly analyzed for certified value and homogeneity.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Specifications
  • MatrixH2O
  • Element/Ion5 SI Units
  • Volume (mL)500
  • Storage SymbolAmbient
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionpH 5 Certified Buffer Standard, 5 SI Units in H2O, 500 mL
Documents & Certificates
Reviews
Q&A