Spex P-GAS-PK Purgeable Aromatics for Gasoline Identification, 2,000 µg/mL; 1 mL; 4/pk

Representative image only.

Spex P-GAS-PK Purgeable Aromatics for Gasoline Identification, 2,000 µg/mL; 1 mL; 4/pk

Spex | Item: 0500892

Analytical standards for municipal and industrial wastewater

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Method 602
  • 2,000 ppm purgeable aromatics for gasoline identification in methanol-P&T matrix
  • Four 1 mL ampules

More About this Item

US EPA Method 602 is a general analytical method for the monitoring of halogenated aromatic compounds in municipal and industrial wastewater by GC/PID.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration1,000 µg/mL (1,000 ppm)
  • MatrixMethanol-P&T
  • ChemistryStandards;Organics
  • Element/IonHeptachlor epoxide (Isomer B)
  • FormLiquid
  • Volume (mL)1
  • Packaging4 x 1 mL
  • DOT InformationHazardous; Methanol; Class 3, 6.1, Packing Group II
  • Storage SymbolAmbient
  • Hazard SymbolFlammable, Acute Toxicity, Irritant, Health Hazard
  • CAS Number1024-57-3
  • UN NumberUN1230
  • Shelf Life (months)36
  • DescriptionHeptachlor Epoxide (Isomer B) Organic Standard, 1,000 µg/mL in Methanol-P&T; 1 mL
Documents & Certificates
Reviews
Q&A