Spex ORG-S8-4Z Sulfur Organometallic Standard, 5,000 µg/mL in Base Oil 75; 50 g

Representative image only.

Spex ORG-S8-4Z Sulfur Organometallic Standard, 5,000 µg/mL in Base Oil 75; 50 g

Spex | Item: 0450112

Analyze for metals in engine oils and other lubricants

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 & ISO 17034:2016
  • Clear transparent mix
  • Track suspended metals to determine the breakdown of specific engine parts
  • 5,000 ppm sulfur in base oil 75
  • 50 g bottle

More About this Item

The determination of wear metals in engine oils and other lubricants can be applied to machines such as automobiles, aircraft, heavy equipment, trucks, locomotives, military vehicles, etc. Specific elements present in used oils have been found to be directly related to specific engine problems. Engine failures, as well as expensive repairs, can be avoided if engine oils are analyzed, providing a periodic trend to predict maintenance or replacement.

Applications
• Wear metals
• Crude oils
• Additive metals
• Environmental monitoring
• Petrochemical testing
• Pharmaceuticals
• Food processing
• Sulfur in diesel fuel

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Concentration5,000 µg/mL (5,000 ppm)
  • MatrixBase Oil 75
  • Element/IonSulfur
  • FormLiquid
  • Weight (g)50
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • UN NumberNot regulated
  • Shelf Life (months)12
  • DescriptionSulfur Organometallic Standard, 5,000 µg/mL in Base Oil 75; 50 g
Documents & Certificates
Reviews
Q&A