Spex ORG-LI8-4Z Lithium Organometallic Standard, 5,000 µg/mL in Base Oil 75; 50 g

Representative image only.

Spex ORG-LI8-4Z Lithium Organometallic Standard, 5,000 µg/mL in Base Oil 75; 50 g

Spex | Item: 0450080

Organometallic oil standards for the determination of metals in oils and lubricants

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Manufactured under an internationally accredited ISO 9001 quality system and compliant with ISO/IEC 17025
  • Guaranteed stable and accurate
  • Clear, transparent mix
  • 5,000 ppm lithium standard in base oil 75 matrix
  • 50 g

More About this Item

The determination of wear metals in engine oils and other lubricants can be applied to machines such as automobiles, aircraft, heavy equipment, trucks, locomotives, military vehicles, etc. The examples are endless. By tracking metals suspended in the used oil, engineers, designers and mechanics can determine the breakdown of specific engine parts. Specific elements present in used oils have been found to be directly related to specific engine problems. Engine failures, as well as expensive repairs, can be avoided if engine oils are analyzed, providing a periodic trend to predict maintenance or replacement.

Applications include:
• Wear metals
• Crude oils
• Additive metals
• Environmental monitoring
• Petrochemical testing
• Pharmaceuticals
• Food processing
• Sulfur in diesel fuel

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Specifications
  • Concentration5,000 µg/mL (5,000 ppm)
  • MatrixBase Oil 75
  • Element/IonLithium (Li)
  • FormLiquid
  • Weight (g)50
  • DOT InformationNot regulated
  • Storage SymbolAmbient
  • Hazard SymbolNot Regulated
  • UN NumberNot regulated
  • Shelf Life (months)12
  • DescriptionLithium Organometallic Standard, 5,000 µg/mL in Base Oil 75; 50 g
Documents & Certificates
Reviews
Q&A