Spex MYCO-1 Mycotoxin Standard Mix, 10 µg/mL in Acetonitrile; 1 mL

Representative image only.

Spex MYCO-1 Mycotoxin Standard Mix, 10 µg/mL in Acetonitrile; 1 mL

Spex | Item: 4105978

Test for mycotoxins to help prevent foodborne illnesses and significant crop losses

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Certified to DQS to ISO 9001:2015
  • Mix includes 5 of the most commonly tested mycotoxins
  • 10 ppm mycotoxin mix in acetonitrile matrix
  • 1 mL ampule

More About this Item

Mold and fungi are all over the world and are of major concern in agricultural products. Fungi and their spores can spread throughout crops and food stores tainting crops. Fungi can proliferate into colonies with the right environmental conditions which destroy crops by consuming them or poison them with toxic secondary metabolites called mycotoxins.

Spex CertiPrep offers an analytical standard mix of five of the most commonly tested mycotoxins (four aflatoxins and ochratoxin A) for your mycotoxin testing needs. Our mycotoxins are ISO 17034 accredited.

Can’t find the standards you are looking for? Spex CertiPrep® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Hazardous material shipping fees may apply.

Specifications
  • Concentration10 µg/mL (10 ppm)
  • MatrixAcetonitrile
  • Element/IonAflatoxin B1, Aflatoxin B2, Aflatoxin G1, Aflatoxin G2, Ochratoxin
  • FormLiquid
  • Volume (mL)1
  • DOT InformationHazardous; Flammable liquids, toxic n.o.s. (Methanol, Acetonitrile); Class 3, 6.1, Packing Group II
  • Storage SymbolFreezer
  • Hazard SymbolFlammable, Toxic, Irritant, Health Hazard
  • CAS Number1162-65-8, 7220-81-7, 1165-39-5, 7241-98-7, 303-47-9
  • UN NumberUN1922
  • DescriptionMycotoxin Standard Mix, 10 µg/mL in Acetonitrile; 1 mL
Documents & Certificates
Reviews
Q&A