Spex MIXSTD2-100 Assurance® Grade Calibration Standard 2 for AA & ICP; 125 mL

Representative image only.

Spex MIXSTD2-100 Assurance® Grade Calibration Standard 2 for AA & ICP; 125 mL

Spex | Item: 0456778

Multi-element calibration standards for AA & ICP instrumentation

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • For use in US EPA Method 200.7 (Revision 4.4) and SW-846, Method 6010 (Third Edition)
  • Mixed multi-element calibration standard 2 for AA & ICP in 5% HNO3 matrix
  • 125 mL bottle

More About this Item

AA & ICP calibration standards are provided for routine instrument calibration. The concentrations and matrices have been selected for convenience of use and stability.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-element calibration standard contains 5 elements.

• Multi-element calibration standard 2 is a 125 mL bottle which contains 10,000 µg/mL (10,000 ppm) Iron and 100 µg/mL (100 ppm) Barium, Cobalt, Copper, Vanadium.

Hazardous material shipping fees may apply.

Specifications
  • ConcentrationMultiple
  • Purity GradeAssurance® Grade
  • Matrix5% HNO3
  • Element/IonBarium, Cobalt, Copper, Iron, and Vanadium
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionAssurance® Grade Mixed Calibration Standard 2 for AA & ICP in 5% HNO3; 125 mL
Documents & Certificates
Reviews
Q&A