Spex MIXSTD1C-500 Assurance® Grade Calibration Standard 1C for AA & ICP; 500 mL

Representative image only.

Spex MIXSTD1C-500 Assurance® Grade Calibration Standard 1C for AA & ICP; 500 mL

Spex | Item: 0456762

Multi-element calibration standards for AA & ICP instrumentation

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • For use in US EPA Method 200.7 (Revision 4.4) and SW-846, Method 6010 (Third Edition)
  • Mixed multi-element calibration standard 1C for AA & ICP in 5% HNO3/tr. Tartaric Acid/tr. HF matrix
  • 500 mL bottle

More About this Item

AA & ICP calibration standards are provided for routine instrument calibration. The concentrations and matrices have been selected for convenience of use and stability.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

This multi-element calibration standard contains 10 elements.

• Multi-element calibration standard 1C is a 500 mL bottle which contains 100 µg/mL (100 ppm) Arsenic, Calcium; 50 µg/mL (50 ppm) Antimony, Selenium; 20 µg/mL (20 ppm) Cadmium, Copper, Manganese; 10 µg/mL (10 ppm) Barium, Boron; and 5 µg/mL (5 ppm) Silver.

Hazardous material shipping fees may apply.

.

Specifications
  • Purity GradeAssurance® Grade
  • ConcentrationMultiple
  • Matrix5% HNO3/tr. Tartaric Acid/tr. HF
  • Element/IonAntimony, Arsenic, Barium, Boron, Cadmium, Calcium, Copper, Manganese, Selenium, and Silver
  • FormLiquid
  • Volume (mL)500
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionAssurance® Grade Mixed Calibration Standard 1C for AA & ICP in 5% HNO3/tr. Tartaric Acid/tr. HF; 500 mL
Documents & Certificates
Reviews
Q&A