Spex MA-CAN-2 Massachusetts Pesticide Residues Mix 2, 100 ppm; 1 mL

Representative image only.

Spex MA-CAN-2 Massachusetts Pesticide Residues Mix 2, 100 ppm; 1 mL

Spex | Item: 4101805

Analytical standards for medicinal and recreational cannabis standards

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Accredited to ISO/IEC 17025:2017 and ISO 17034:2016

More About this Item

With the legalization of cannabis, for both medicinal and recreational purposes, has been gaining speed, legislation and regulation has not necessarily kept pace. Even so, out of a drive for self-regulation and significant consumer safety concerns, many producers and manufacturers are turning to testing labs in order to ensure that their products are high quality and free of chemical contaminants. Spex CertiPrep offers ISO 17034 Certified Reference Materials (CRMs) for all of the common contaminants such as pesticide residues, residual solvents and heavy metals, as well as qualitative analysis CRMs such as terpenes. As the industry demands change and regulations are put into place, we continually update our product offerings.

Designed for state specific pesticide regulations:
OAR 333-008-11
HB 3460
AOAC 2007-01
EN 15662

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CannStandards® can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
Hazardous material shipping fees may apply.

SPEX SamplePrep does not support or promote the use of its products or services in connection with illegal cultivation of cannabis products. SPEX SamplePrep products are intended to be used only in states where the cultivation, sale and use of cannabis products are legal.

Specifications
  • Concentration100 µg/mL (100 ppm)
  • MatrixLC/MS Acetonitrile
  • Element/IonAbamectin, Azadirachtin, Azoxystrobin, Boscalid, Carbaryl, Chlorfenapyr, Diotefuran, Imbda-Cyhalothrin, Paclobutrazol, Permethrin, Piperonyl butoxide, Myrethrins (mix of isomers), Spinosad, Spirotetramat
  • Volume (mL)1
  • Storage SymbolAmbient
  • CAS Number71751-41-2, 11141-17-6, 131860-33-8, 188425-85-6, 63-25-2, 122453-73-0, 165252-70-0, 91465-08-6, 76738-62-0, 52645-53-1, 51-03-6, 8003-34-7, 168316-95-8, 203313-25-1
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionMassachusetts Pesticide Residues Mix 2, 100 ppm; 1 mL
Documents & Certificates
Reviews
Q&A