Spex LPC-1-100N Assurance® Grade LPC Standard without Mercury for AA & ICP in 5% HNO3/tr. Tartaric Acid/tr. HF, 125 mL

Representative image only.

Spex LPC-1-100N Assurance® Grade LPC Standard without Mercury for AA & ICP in 5% HNO3/tr. Tartaric Acid/tr. HF, 125 mL

Spex | Item: 0456817

Mixed multi-element standard for AA and ICP

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Diluted in 5% HNO3/tr. Tartaric Acid/tr. HF
  • Use standard to check spectral interferences

More About this Item

Assurance® Grade standards may be diluted in the same matrix as specified; however, caution must be exercised in the choice of the source for your diluents. Diluting the matrix may cause some standards to precipitate. Also, an impure or unknown diluent turns your standard into an unknown. We recommend using only Spex CertiPrep® Matrix Blanks when diluting your standards.

The Laboratory Performance Check (LPC) standard is a solution of method analysis analytes used to evaluate the performance of the instrument. The LPC standard is used immediately following calibration, after every tenth sample, and at the end of the sample run. The analyzed value of each analyte in the LPC solution should be within 95% to 105% of its expected value. If the analyte value is outside of the interval, reanalyze the LPC. If the analyte is again outside of the +/- 5% limit, the instrument should be recalibrated and all samples following the last acceptable LPC solution should be reanalyzed.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

Hazardous material shipping fees may apply.

Specifications
  • Purity GradeAssurance® Grade
  • Matrix5% HNO3/tr. Tartaric Acid/tr. HF
  • ClassAA & ICP
  • Element/IonPhosphorus, Potassium, Silica, Aluminum, Antimony, Arsenic, Barium, Beryllium, Boron, Cadmium, Calcium, Chromium, Cobalt, Copper, Iron, Lead, Lithium, Magnesium, Manganese, Molybdenum, Nickel, Selenium, Sodium, Strontium, Thallium, Tin, Vanadium, Zinc, Silver
  • FormLiquid
  • Volume (mL)125
  • DOT InformationHazardous
  • Storage SymbolAmbient
  • UN NumberUN3264
  • Shelf Life (days)365
  • Shelf Life (months)12
  • DescriptionAssurance® Grade LPC Standard without Mercury for AA & ICP in 5% HNO3/tr. Tartaric Acid/tr. HF, 125 mL
Documents & Certificates
Reviews
Q&A