Spex LFS-1-500N Assurance® Grade LFS Solution 1 without Mercury for AA & ICP in 5% HNO3/tr. Tartaric Acid/tr. HF; 500 mL

Representative image only.

Spex LFS-1-500N Assurance® Grade LFS Solution 1 without Mercury for AA & ICP in 5% HNO3/tr. Tartaric Acid/tr. HF; 500 mL

Spex | Item: 0456813

Multi-element standards for AA & ICP analysis

Call For Price

Call For Price
Product Description

Includes

Supplier Certificate of Analysis (CoA) is included with product shipment.

Key Features

  • Spike laboratory fortified blank and sample matrix
  • Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
  • Laboratory fortifying stock 1 without mercury in 5% HNO3/tr. tartaric acid/tr. HF matrix
  • 500 mL bottle

More About this Item

Laboratory Fortifying Stock (LFS) Solutions is used for spiking the laboratory fortified blank and the laboratory fortified sample matrix. Two (2 mL) of the LFS solution must be added to a 100 mL aliquot of the laboratory fortified blank. This blank must be carried through the entire sample preparation procedure and analysis scheme.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.

This multi-element solution contains 25 components.

• Laboratory fortifying solution (LFS) 1 without mercury is a 500 mL bottle which contains 50 µg/mL (50 ppm) of Phosphorus; 25 µg/mL (25 ppm) of Aluminum, Antimony, Arsenic, Barium, Boron, Chromium, Copper, Iron, Lead, Lithium, Manganese, Nickel, Selenium, Silica, Strontium, Thallium, Zinc; 10 µg/mL (10 ppm) of Cadmium, Cobalt, Molybdenum, Tin, Vanadium; 5 µg/mL (5 ppm) of Beryllium; and 2.5 µg/mL (2.5 ppm) of SIlver.

Hazardous material shipping fees may apply.

LFS Solution 1 does not contain Ca, K, Mg, or Na because their concentration will vary from one environmental sample to the other.

Specifications
  • Purity GradeAssurance® Grade
  • ConcentrationMultiple
  • Matrix5% HNO3/tr. Tartaric Acid/tr. HF
  • Element/IonMultiple
  • FormLiquid
  • Volume (mL)500
  • DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
  • Storage SymbolAmbient
  • Hazard SymbolCorrosion
  • UN NumberUN3264
  • Shelf Life (months)12
  • DescriptionAssurance® Grade LFS Solution 1 without Mercury for AA & ICP in 5% HNO3/tr. Tartaric Acid/tr. HF; 500 mL
Documents & Certificates
Reviews
Q&A