Supplier Certificate of Analysis (CoA) is included with product shipment.
Key Features
Spike laboratory fortified blank and sample matrix
Manufactured under ISO 9001, ISO/IEC 17025 and ISO 17034 compliance
Laboratory fortifying stock 1 with mercury in 5% HNO3/tr. tartaric acid/tr. HF matrix
125 mL bottle
More About this Item
Laboratory Fortifying Stock (LFS) Solutions is used for spiking the laboratory fortified blank and the laboratory fortified sample matrix. Two (2 mL) of the LFS solution must be added to a 100 mL aliquot of the laboratory fortified blank. This blank must be carried through the entire sample preparation procedure and analysis scheme.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Can’t find the standards you are looking for? Spex CertiPrep®can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 732-549-7144.
This multi-element solution contains 2 solutions.
• Solution 1 is a 125 mL bottle which contains 50 µg/mL (50 ppm) of Phosphorus; 25 µg/mL (25 ppm) of Aluminum, Antimony, Arsenic, Barium, Boron, Chromium, Copper, Iron, Lead, Lithium, Manganese, Nickel, Selenium, Silica, Strontium, Thallium, Zinc; 10 µg/mL (10 ppm) of Cadmium, Cobalt, Molybdenum, Tin, Vanadium; 5 µg/mL (5 ppm) of Beryllium; and 2.5 µg/mL (2.5 ppm) of SIlver in 5% HNO3/tr. Tartaric Acid/tr. HF. This is available individually as part number 04568-12.
• Solution 2 is a 125 mL bottle which contains 10 µg/mL (10 ppm) of Mercury in 5% HNO3. This is available individually as part number 04503-58.
Hazardous material shipping fees may apply.
LFS Solution 1 does not contain Ca, K, Mg, or Na because their concentration will vary from one environmental sample to the other.
Specifications
ConcentrationMultiple
Purity GradeAssurance® Grade
Matrix5% HNO3/tr. Tartaric Acid/tr. HF
Element/IonTwo bottle set
FormLiquid
Volume (mL)125
DOT InformationHazardous; Corrosive liquid, acidic, inorganic, n.o.s, (Nitric acid solution); Class 8, Packing Group III
Storage SymbolAmbient
Hazard SymbolCorrosion
UN NumberUN3264
Shelf Life (months)12
DescriptionAssurance® Grade LFS Solution 1 with Mercury for AA & ICP in 5% HNO3/tr. Tartaric Acid/tr. HF; 125 mL